missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Secondary Vacuum Traps

For any mechanically refrigerated freeze-dry system; ensure efficient processing of samples with low eutectic points

Supplier:  Labconco™ 7538000

 View more versions of this product

Catalog No. 1026946A


Only null left

Requires : To find a pump for use with this trap, see Pumps.

Description

Description

For any mechanically refrigerated freeze-dry system; ensure efficient processing of samples with low eutectic points

  • Trap will cool to approximately -75°C (-103°F) with dry ice and solvent in its removable well
  • Can be used as an inexpensive collector: connect a sample to incoming port, vacuum pump to outgoing port, and system is complete
  • Stainless-steel, with cover

Specifications

Chemical Identifiers

Stainless Steel
Refrigerated freeze-dry system
−75°C
Prevent contaminants from migrating into the vacuum pump
−103°F

Specifications

0.237 in.
0.75 in.
900 mL
Prevent contaminants from migrating into the vacuum pump
9.75 in.
0.75 in. o.d. inlet/outlet connection
−75°C
3100 mL
4.1 kg
900 mL
1.9 cm
Stainless Steel
Refrigerated freeze-dry system
24.76 cm
−103°F
0.818 gal.
9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions