missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Secnidazole | |
| 74°C | |
| C7H11N3O3 | |
| 1 g | |
| KPQZUUQMTUIKBP-UHFFFAOYSA-N | |
| 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol | |
| 71815 | |
| ≥98.0% (GC) |
| 3366-95-8 | |
| White-Yellow | |
| MFCD00864656 | |
| 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol | |
| CC1=NC=C(N1CC(C)O)[N+](=O)[O-] | |
| 185.183 | |
| 185.18 | |
| Crystalline Powder |
Chemical Identifiers
| 3366-95-8 | |
| 185.183 | |
| KPQZUUQMTUIKBP-UHFFFAOYSA-N | |
| 71815 | |
| CC1=NC=C(N1CC(C)O)[N+](=O)[O-] |
| C7H11N3O3 | |
| MFCD00864656 | |
| 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol | |
| 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol |
Safety and Handling
TSCA : No