missing translation for 'onlineSavingsMsg'
Learn More

Secnidazole 98.0+%, TCI America™

Catalog No. S09115G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09115G 5 g
S09111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09115G Supplier TCI America Supplier No. S09115G
Only null left

Chemical Identifiers

CAS 3366-95-8
Molecular Formula C7H11N3O3
Molecular Weight (g/mol) 185.183
MDL Number MFCD00864656
InChI Key KPQZUUQMTUIKBP-UHFFFAOYSA-N
Synonym 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol
PubChem CID 71815
IUPAC Name 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
SMILES CC1=NC=C(N1CC(C)O)[N+](=O)[O-]

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 185.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Secnidazole