missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Secador™ 5.0 Desiccator Cabinet

Catalog No. 01257489
Change view
Click to view available options
No. of Shelves:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Shelves
01257489 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257489 Supplier Bel-Art™ Supplier No. H420570001
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cabinet provides dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament.

  • High-density foam gaskets to keep ambient humidity out and dry air inside
  • Large clear polycarbonate panels are durable and allow for easy viewing of internal contents
  • LCD shows the internal temperature and humidity level
  • Includes 3 adjustable and removable shelves

Specifications

Autoclavable Not Autoclavable
Color White
Cabinet Style Front-loading
Material Polycarbonate
Product Type Desiccator
Door Style Hinged
No. of Shelves 3
No. per Pack 1 Ea.
Shelf Positions Adjustable
Dimensions (D x W x H) Interior 34 x 28 x 50 cm
Width (English) 11.8 in.
Width (Metric) 30 cm
Height (English) 20.4 in.
Height (Metric) 52 cm
Depth (English) 13 in.
Depth (Metric) 34 cm
Show More Show Less