missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Secador™ 5.0 Desiccator Cabinet PROMO
Cabinet provides dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament.
Supplier: Bel-Art™ H420570001PM
Description
- High-density foam gaskets to keep ambient humidity out and dry air inside
- Large clear polycarbonate panels are durable and allow for easy viewing of internal contents
- LCD shows the internal temperature and humidity level
- Includes 3 adjustable and removable shelves
Cabinet, dry storage, storage cabinet
Specifications
| Not Autoclavable | |
| Front-loading | |
| Desiccator | |
| 2 | |
| 34 x 28 x 50 cm | |
| 30 cm | |
| 52 cm | |
| 34 cm |
| White | |
| Polycarbonate | |
| Hinged | |
| Adjustable | |
| 11.8 in. | |
| 20.4 in. | |
| 13 in. |