missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Secador™ 5.0 Adjustable Auto-Desiccator Cabinet, Electric Non-Desiccant PROMO

Catalog No. 01257492PM
Change view
Click to view available options
No. of Shelves:
3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Shelves
01257492PM 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257492PM Supplier Bel-Art™ Supplier No. H420571004PM

Cabinet provides dry storage for humidity-sensitive products such as laboratory samples, reagents, optical equipment, electronics, and 3D printing filament. Can be programmed to desired humidity level.

  • High-density foam gaskets to keep ambient humidity out and dry air inside
  • Large clear polycarbonate panels are durable and allow for easy viewing of internal contents
  • LCD shows the internal temperature and humidity level
  • Includes 3 adjustable and removable shelves
  • Electric models keeps internal humidity at 20 to 40%

Cabinet, dry storage, storage cabinet

Specifications

Autoclavable Not Autoclavable
Color White
Cabinet Style Front-loading
Material Polycarbonate
Product Type Desiccator
Door Style Hinged
No. of Shelves 3
Shelf Positions Adjustable
Dimensions (D x W x H) Interior 34 x 28 x 50 cm
Width (English) 11.8 in.
Width (Metric) 30 cm
Height (English) 25 in.
Height (Metric) 63.5 cm
Depth (English) 13 in.
Depth (Metric) 34 cm
Voltage 100/120 V or 200/240 V
Show More Show Less