missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58714AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58714AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58714AD Supplier Thermo Scientific Chemicals Supplier No. H58714AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 171860-66-5 | C4H9BrZn | 202.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent

Chemical Identifiers

CAS 171860-66-5
Molecular Formula C4H9BrZn
Molecular Weight (g/mol) 202.4
MDL Number MFCD00671990
InChI Key DATGUFISTUEOBS-UHFFFAOYSA-M
Synonym 2-butylzinc bromide, bromo sec-butyl zinc, zinc, bromo 1-methylpropyl, sec-butylzinc bromide, 0.50 m in thf, 2-butylzinc bromide 0.5 m in tetrahydrofuran, sec-butylzinc bromide solution, 0.5 m in thf, sec-butylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 4101435
IUPAC Name bromozinc(1+);butane
SMILES CC[CH-]C.[Zn+]Br

Specifications

Density 0.968
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 202.4
Chemical Name or Material sec-Butylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only