missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylamine 99.0+%, TCI America™

Catalog No. p-7118626
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B070825ML 25 mL
B0708500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B070825ML Supplier TCI America Supplier No. B070825ML
Only null left

Chemical Identifiers

CAS 13952-84-6
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008094
InChI Key BHRZNVHARXXAHW-UHFFFAOYNA-N
Synonym sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote
PubChem CID 24874
ChEBI CHEBI:74526
IUPAC Name butan-2-amine
SMILES CCC(C)N

Specifications

Color Colorless
Boiling Point 63°C
Quantity 25 mL
UN Number 2733
Formula Weight 73.14
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material sec-Butylamine
EINECSNumber (2)-0132
RTECSNumber EO3325000
TSCA Yes