missing translation for 'onlineSavingsMsg'
Learn More

sec-Butylamine, 99%

Catalog No. AC107810050
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107810050 5 mL Glass bottle
AC107811000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107810050 Supplier Thermo Scientific Chemicals Supplier No. 107810050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13952-84-6 | C4H11N | 73.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13952-84-6
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008094
InChI Key BHRZNVHARXXAHW-UHFFFAOYNA-N
Synonym sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote
PubChem CID 24874
ChEBI CHEBI:74526
IUPAC Name butan-2-amine
SMILES CCC(C)N

Specifications

Melting Point -104.0°C
Color Colorless to Yellow
Density 0.7200g/mL
Boiling Point 63.0°C
Flash Point −19°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3920 to 1.3960
Linear Formula C2H5CH(NH2)CH3
Quantity 5 mL
Beilstein 04,16
Merck Index 15,1546
Specific Gravity 0.72
Solubility Information Solubility in water: miscible. Other solubilities: freely miscible with most common organic solvents
Viscosity 0.5 mPa.s (20°C)
Formula Weight 73.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material sec-Butylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 237-732-7

RUO – Research Use Only