missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl methyl ether, 99%

Catalog No. p-7035703
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53427AA 5 mL
AAH53427AC 25 mL
AAH53427AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH53427AA Supplier Thermo Scientific Chemicals Supplier No. H53427AA

CAS: 6795-87-5 | C5H12O | 88.15 g/mol

sec-Butyl methyl ether is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl methyl ether is used as a solvent in organic synthesis.

Solubility
Miscible with water, alcohol, ether and acetone.

Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.

Chemical Identifiers

CAS 6795-87-5
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00190205
InChI Key FVNIMHIOIXPIQT-UHFFFAOYNA-N
Synonym sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a
PubChem CID 23238
IUPAC Name 2-methoxybutane
SMILES CCC(C)OC

Specifications

Density 0.742
Boiling Point 57°C
Flash Point −30°C (−22°F)
Refractive Index 1.372
Quantity 5 mL
UN Number UN2350
Solubility Information Miscible with water,alcohol,ether and acetone.
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material sec-Butyl methyl ether
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER
RTECSNumber KN5249500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only