missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Ether (DL- and meso- mixture) (stabilized with HQ) 97.0+%, TCI America™
$117.43 - $352.30
Chemical Identifiers
| CAS | 6863-58-7 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.231 |
| MDL Number | MFCD00039921 |
| InChI Key | HHBZZTKMMLDNDN-UHFFFAOYSA-N |
| Synonym | Di-sec-butyl Ether |
| PubChem CID | 23294 |
| IUPAC Name | 2-butan-2-yloxybutane |
| SMILES | CCC(C)OC(C)CC |
Chemical Identifiers
| 6863-58-7 | |
| 130.231 | |
| HHBZZTKMMLDNDN-UHFFFAOYSA-N | |
| 23294 | |
| CCC(C)OC(C)CC |
| C8H18O | |
| MFCD00039921 | |
| Di-sec-butyl Ether | |
| 2-butan-2-yloxybutane |
Specifications
| 6863-58-7 | |
| 121°C | |
| MFCD00039921 | |
| 1149 | |
| HHBZZTKMMLDNDN-UHFFFAOYSA-N | |
| 2-butan-2-yloxybutane | |
| 23294 | |
| ≥97.0% (GC) | |
| sec-Butyl Ether (DL- and meso- mixture) (stabilized with HQ) |
| Colorless | |
| C8H18O | |
| 5 mL | |
| Di-sec-butyl Ether | |
| CCC(C)OC(C)CC | |
| 130.231 | |
| 130.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0363
RTECSNumber : EQ2220000
TSCA : Yes