missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Acetate 98.0+%, TCI America™

Catalog No. p-7116666
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A002525ML 25 mL
A0025500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A002525ML Supplier TCI America Supplier No. A002525ML

Chemical Identifiers

CAS 105-46-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009328
InChI Key DCKVNWZUADLDEH-UHFFFAOYNA-N
Synonym sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate
PubChem CID 7758
IUPAC Name butan-2-yl acetate
SMILES CCC(C)OC(C)=O

Specifications

Color Colorless
Boiling Point 112°C
Quantity 25 mL
UN Number 1123
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material sec-Butyl Acetate
EINECSNumber (2)-0731
RTECSNumber AF7380000
TSCA Yes