missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sec-Butyl Acetate 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 112°C |
| Quantity | 500 mL |
| UN Number | 1123 |
| Formula Weight | 116.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | sec-Butyl Acetate |
Safety and Handling
| EINECSNumber | (2)-0731 |
| RTECSNumber | AF7380000 |
| TSCA | Yes |