missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl acetate, 98%

Catalog No. AAL1389214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1389214 25 g
AAL1389222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1389214 Supplier Thermo Scientific Chemicals Supplier No. L1389214

CAS: 105-46-4 | C6H12O2 | 116.16 g/mol

sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl acetate is used in organic synthesis, catalytic agent, petrochemical additive.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 105-46-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009328
InChI Key DCKVNWZUADLDEH-UHFFFAOYNA-N
Synonym sec-butyl acetate, dl-sec-butyl acetate, 2-butyl acetate, acetic acid, 1-methylpropyl ester, 1-methylpropyl acetate, sec-butyl ethanoate, 1-methylpropyl ethanoate, acetic acid, sec-butyl ester, 2-butanol acetate, sec-butyl alcohol acetate
PubChem CID 7758
IUPAC Name butan-2-yl acetate
SMILES CCC(C)OC(C)=O

Specifications

Melting Point -99°C
Density 0.872
Boiling Point 111°C to 112°C
Flash Point 16°C (60°F)
Refractive Index 1.389
Quantity 25 g
UN Number UN1123
Beilstein 1720689
Merck Index 14,1536
Solubility Information Slightly soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material sec-Butyl acetate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-300-1
RTECSNumber AF7380000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only