Learn More
sec-Butanol, 99%, Extra Pure
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Supplier: Thermo Scientific Chemicals 107700025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| sec-Butanol | |
| 78-92-2 | |
| 99.0°C | |
| 24°C | |
| Glass Bottle | |
| 1.3960 to 1.3980 (20°C, 589nm) | |
| 99% | |
| C4H10O | |
| MFCD00004569 | |
| 0.5% max. | |
| 15, 1543 | |
| Solubility in water: 125g/L (20°C). Other solubilities: soluble in alcohol, ester, ether and benzene | |
| CCC(C)O | |
| 74.12 | |
| CHEBI:35687 | |
| 99% |
| -115.0°C | |
| 0.8100g/mL | |
| 2.5 L | |
| Conforms to Ref Report | |
| Liquid | |
| 0.005% max. | |
| 0.81 | |
| C2H5CH(OH)CH3 | |
| 4.2 mPa.s (20°C) | |
| 01, 371 | |
| 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
| BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
| butan-2-ol | |
| 6568 | |
| 74.12 | |
| Extra Pure |
Chemical Identifiers
| 78-92-2 | |
| 74.12 | |
| BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
| 6568 | |
| butan-2-ol |
| C4H10O | |
| MFCD00004569 | |
| 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
| CHEBI:35687 | |
| CCC(C)O |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only