missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl Chloride 95.0+%, TCI America™

Catalog No. p-7135773
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S003025ML 25 mL
S0030100ML 100 mL
S0030500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S003025ML Supplier TCI America Supplier No. S003025ML
Only null left

Chemical Identifiers

CAS 111-19-3
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.136
MDL Number MFCD00000770
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Melting Point -3°C
Color Brown-Yellow
Boiling Point 167°C
Quantity 25 mL
UN Number 2922
Formula Weight 239.14
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Sebacoyl Chloride
EINECSNumber (2)-2616
RTECSNumber HD8454250
TSCA Yes
Recommended Storage Refrigerator