missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl chloride, 95%

Catalog No. AAA1422636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1422636 500 g
AAA1422614 25 g
AAA1422622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1422636 Supplier Thermo Scientific Chemicals Supplier No. A1422636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111-19-3 | C10H16Cl2O2 | 239.136 g/mol

Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

Solubility
Miscible with hydrocarbons and ethers.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.

Chemical Identifiers

CAS 111-19-3
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.136
MDL Number MFCD00000770
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Melting Point -5°C
Density 1.121
Boiling Point 161°C (14 mmHg)
Flash Point 165°C (329°F)
Odor Acrid
Refractive Index 1.468
Quantity 500 g
UN Number UN2922
Beilstein 1365665
Sensitivity Moisture sensitive
Solubility Information Miscible with hydrocarbons and ethers.
Formula Weight 239.14
Percent Purity 97%
Chemical Name or Material Sebacoyl chloride
Show More Show Less
Hazard Category H302-H310-H314-H335
Hazard Statement GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-843-4
RTECSNumber HD8454250
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only