missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl chloride, 92%, technical

Catalog No. p-191879
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294900500 50 g Glass bottle
AC294902500 250 g Glass bottle
AC294905000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC294900500 Supplier Thermo Scientific Chemicals Supplier No. 294900500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC) | CAS: 111-19-3 | C10H16Cl2O2 | 239.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-19-3
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.14
MDL Number MFCD00000770
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Melting Point -2.5°C
Color White to Beige
Density 1.1210g/mL
Boiling Point 220.0°C (75.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Refractive Index 1.4670 to 1.4700
Linear Formula ClCO(CH2)8COCl
Quantity 50 g
Beilstein 02, 719
Specific Gravity 1.121
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in hydrocarbons and ethers
Formula Weight 239.14
Percent Purity 92%
Grade Technical
Physical Form Liquid
Chemical Name or Material Sebacoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath
EINECSNumber 203-843-4
RTECSNumber HD8454250
TSCA TSCA
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only