missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sebaconitrile 98.0+%, TCI America™
$125.13 - $1004.93
Chemical Identifiers
| CAS | 1871-96-1 |
|---|---|
| Molecular Formula | C10H16N2 |
| Molecular Weight (g/mol) | 164.252 |
| MDL Number | MFCD00019900 |
| InChI Key | DFJYZCUIKPGCSG-UHFFFAOYSA-N |
| Synonym | sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference |
| PubChem CID | 74639 |
| IUPAC Name | decanedinitrile |
| SMILES | C(CCCCC#N)CCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S002925ML
|
TCI America
S002925ML |
25 mL |
Each for $125.13
|
|
|||||
|
S0029500ML
|
TCI America
S0029500ML |
500 mL |
Each for $1,004.93
|
|
|||||
Chemical Identifiers
| 1871-96-1 | |
| 164.252 | |
| DFJYZCUIKPGCSG-UHFFFAOYSA-N | |
| 74639 | |
| C(CCCCC#N)CCCC#N |
| C10H16N2 | |
| MFCD00019900 | |
| sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference | |
| decanedinitrile |
Specifications
| 1871-96-1 | |
| 200°C | |
| MFCD00019900 | |
| 3276 | |
| DFJYZCUIKPGCSG-UHFFFAOYSA-N | |
| decanedinitrile | |
| 74639 | |
| ≥98.0% (GC) | |
| Sebaconitrile |
| Yellow | |
| C10H16N2 | |
| 25 mL | |
| sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference | |
| C(CCCCC#N)CCCC#N | |
| 164.252 | |
| 164.25 | |
| Liquid |
Safety and Handling
RTECSNumber : VS1500000
TSCA : Yes