missing translation for 'onlineSavingsMsg'
Learn More

Sebaconitrile 98.0+%, TCI America™

Catalog No. S002925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S002925ML 25 mL
S0029500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S002925ML Supplier TCI America Supplier No. S002925ML

Chemical Identifiers

CAS 1871-96-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00019900
InChI Key DFJYZCUIKPGCSG-UHFFFAOYSA-N
Synonym sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference
PubChem CID 74639
IUPAC Name decanedinitrile
SMILES C(CCCCC#N)CCCC#N

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 mL
UN Number 3276
Formula Weight 164.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Sebaconitrile
RTECSNumber VS1500000
TSCA Yes