missing translation for 'onlineSavingsMsg'
Learn More

Sebaconitrile, 98%

Catalog No. AC419250250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419250250 25 g Glass bottle
AC419251000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC419250250 Supplier Thermo Scientific Chemicals Supplier No. 419250250

CAS: 1871-96-1 | C10H16N2 | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1871-96-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.25
MDL Number MFCD00019900
InChI Key DFJYZCUIKPGCSG-UHFFFAOYSA-N
Synonym sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference
PubChem CID 74639
IUPAC Name decanedinitrile
SMILES C(CCCCC#N)CCCC#N

Specifications

Melting Point 7.0°C
Color Colorless to Yellow
Density 0.9100g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490
Linear Formula NC(CH2)8CN
Quantity 25 g
Specific Gravity 0.91
Formula Weight 164.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Sebaconitrile, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
EINECSNumber 217-494-0
RTECSNumber VS1500000
TSCA TSCA

RUO – Research Use Only