Learn More
Sebaconitrile, 95%
CAS: 1871-96-1 | C10H16N2 | 164.252 g/mol
$50.67 - $161.70
Chemical Identifiers
| CAS | 1871-96-1 |
|---|---|
| Molecular Formula | C10H16N2 |
| Molecular Weight (g/mol) | 164.252 |
| MDL Number | MFCD00019900 |
| InChI Key | DFJYZCUIKPGCSG-UHFFFAOYSA-N |
| Synonym | sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference |
| PubChem CID | 74639 |
| IUPAC Name | decanedinitrile |
| SMILES | C(CCCCC#N)CCCC#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2301006
|
Thermo Scientific Chemicals
B2301006 |
5 g |
N/A
|
|
|||||
|
AAB2301014
|
Thermo Scientific Chemicals
B2301014 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1871-96-1 | |
| 164.252 | |
| DFJYZCUIKPGCSG-UHFFFAOYSA-N | |
| 74639 | |
| C(CCCCC#N)CCCC#N |
| C10H16N2 | |
| MFCD00019900 | |
| sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference | |
| decanedinitrile |
Specifications
| 1871-96-1 | |
| 0.91 | |
| >110°C (230°F) | |
| MFCD00019900 | |
| 1750739 | |
| DFJYZCUIKPGCSG-UHFFFAOYSA-N | |
| decanedinitrile | |
| 74639 | |
| 95% |
| 4°C to 8°C | |
| 199°C to 200°C | |
| C10H16N2 | |
| 5 g | |
| sebaconitrile, 1,8-dicyanooctane, octamethylene dicyanide, oktamethylendikyanid, oktamethylendikyanid czech, octane-1,8-dicarbonitrile, sebacic acid dinitrile, acmc-209epg, wln: nc8cn, 4-02-00-02089 beilstein handbook reference | |
| C(CCCCC#N)CCCC#N | |
| 164.252 | |
| 164.25 | |
| Sebaconitrile |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 217-494-0
RTECSNumber : VS1500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only