missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sebacic Dihydrazide 96.0+%, TCI America™
$109.98 - $749.35
Chemical Identifiers
| CAS | 925-83-7 |
|---|---|
| Molecular Formula | C10H22N4O2 |
| Molecular Weight (g/mol) | 230.312 |
| MDL Number | MFCD00039763 |
| InChI Key | ZWLIYXJBOIDXLL-UHFFFAOYSA-N |
| Synonym | Decanediohydrazide, Sebacohydrazide |
| PubChem CID | 70219 |
| IUPAC Name | decanedihydrazide |
| SMILES | C(CCCCC(=O)NN)CCCC(=O)NN |
Chemical Identifiers
| 925-83-7 | |
| 230.312 | |
| ZWLIYXJBOIDXLL-UHFFFAOYSA-N | |
| 70219 | |
| C(CCCCC(=O)NN)CCCC(=O)NN |
| C10H22N4O2 | |
| MFCD00039763 | |
| Decanediohydrazide, Sebacohydrazide | |
| decanedihydrazide |
Specifications
| 925-83-7 | |
| White | |
| MFCD00039763 | |
| Decanediohydrazide, Sebacohydrazide | |
| C(CCCCC(=O)NN)CCCC(=O)NN | |
| 230.312 | |
| 230.31 | |
| Crystalline Powder |
| 186°C | |
| C10H22N4O2 | |
| 25 g | |
| ZWLIYXJBOIDXLL-UHFFFAOYSA-N | |
| decanedihydrazide | |
| 70219 | |
| ≥96.0% (T) | |
| Sebacic Dihydrazide |
Safety and Handling
EINECSNumber : (2)-0906
RTECSNumber : VS1225000
TSCA : Yes