missing translation for 'onlineSavingsMsg'
Learn More

Calcium Phosphate Dibasic, Anhydrous, Powder, FCC, EP, BP, 98-101%, Spectrum™ Chemical

Catalog No. 18601347
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601347 2.5 kg Poly Bottle
18601346 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18601347 Supplier Spectrum Chemical Mfg Cor Supplier No. C11512.5KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1151, 7757-93-9, CaHPO4

Spectrum™ Chemical Calcium Phosphate Dibasic, Anhydrous, Powder, FCC, EP, BP is used as a calcium dietary supplement in prepared breakfast cereals, dog treats, enriched flour, and noodle products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
IUPAC Name calcium hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Physical Form Powder, Needles or Crystals
CAS Max % 100%
Quantity 2.5 kg
Loss on Ignition 7.0 to 8.5%
Packaging Poly Bottle
Percent Purity 98 to 101%
Grade Multi-Compendial
Chemical Name or Material Calcium Phosphate, Dibasic, Anhydrous