Learn More
Search Latent Print Value Kit
Catalog No. S69025A
Change view
Click to view available options
Latent Print Value Kit
Catalog No.
S69025A Latent Print Value Kit
1 options
Components in this kit contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of this product.
Catalog No. S69025A Supplier Customer Channels Group (Ccg) Supplier No. 161LV
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Components in this kit contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of this product.
Includes
One 1.5 x 60 in. clear lift tape, 20 3 x 5 in. white backing/sketch cards, one regular powder brush, 1 oz. black latent powder, one pair disposable latex gloves, one molded polypropylene carrying case
Economical kit for crime scene investigation.
Everything you need to recover fingerprints at an affordable cost.
Specifications
For Use With (Application) For crime scene investigation
Includes One 144l 1.5 in. clear lift tape, 20 lpw1001 3 x 5 in. backing/sketch cards, white, one 118l regular powder brush, one 101l black latent powder, 1 oz, 1 sf00771 disposable latex gloves, pair, 1 molded, polypropylene carrying case
Product Type Latent Print Value Kit