missing translation for 'onlineSavingsMsg'
Learn More

PIP™ Polyurethane Coated Seamless Knit Polyester Gloves

Catalog No. 19046394
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19046394 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19046394 Supplier PIP™ Supplier No. GP33G115M

Seamless knit polyester shell offers increased comfort, finger dexterity, and breathability. Polyurethane coated flat grip on the palm and fingers provides tactility in dry and slightly oily conditions.

  • Seamless knit polyester shell increases comfort, finger dexterity, and breathability
  • Polyurethane coating takes on the surface properties of the glove liner to provide a tactile grip in dry and slightly oily conditions
  • Ideal for applications that require dexterity for precise handling
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Breathable back for comfort

Specifications

ANSI Abrasion Rating ANSI 2
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Exterior Finish No Texture
Hand Specific
Material Polyester
Product Type Seamless Knit Gloves
Size Medium
Style Standard
For Use With (Application) Electronics Assembly, Inspection, and Quality Control
Glove Cuff Style Knit
Series 33-G115
Yarn Gauge 13
Length (English) 9.4 in.
Length (Metric) 24 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov