Learn More
Alkali Scientific SealSafe Titan™ Sealing Film Rolls, 2 Rolls/pk, 100 Sheets/Roll
Catalog No. 502389331
Encompass
SealSafe's dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded green band. (This item is for 2 replacement rolls only. For stater kit, order SKU SF25ML or SF25ML-S for sterile version.)
Catalog No. 502389331 Supplier Alkali Scientific Supplier No. SF24ML
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SealSafe's dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Environmentally friendly; no paper liner to remove and discard; utilizes approximately 72% less material than SealPlate sheeted film. Each sheet is separated by an adhesive-free, perforated, color-coded green band. (This item is for 2 replacement rolls only. For stater kit, order SKU SF25ML or SF25ML-S for sterile version.)