missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ampac™ SealPAK Tubullar Rollstock Pouches
Create custom-size pouches
Supplier: Ampac™ TRSF951253
Includes: Pouch cutter (01-812-21) supplied with every roll except 01-812-26A.
Description
Create custom-size pouches
- 3-ply laminated structure
- Composed of polyester, foil and polypropylene (PET/Foil/PP)
- Wthstands temperatures up to 130°C (266°F)
- Can be used in autoclave applications with over-pressure at cool down
- Offers oxygen and moisture barriers
Compliance
Use no adhesives or bonding agents.
Chemical Identifiers
| Heat-Sealable Roll Stock Pouches |
Specifications
| Heat-Sealable Roll Stock Pouches | |
| 4.50 mil | |
| 125 ft. | |
| 38 m |
| Foil | |
| 125 ft. x 9.5 in. | |
| 9.5 in. | |
| 24 cm |