Learn More
Alkali Scientific ASI™ 96 Well Round Silicone Sealing Mat, Pre-slit, Piercable, 1.0mL, 10/pk, 50/cs
Catalog No. 502336005
Encompass
ASI™ 96 Well Silicone Sealing Mat, 50/case are designed for 96-well storage plates and are available in piercable format. The square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.
Catalog No. 502336005 Supplier Alkali Scientific Supplier No. MPD51S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ASI™ 96 Well Silicone Sealing Mat, 50/case are designed for 96-well storage plates and are available in piercable format. The square capped mats are made of thermoplastic elastomer and are suitable for long-term storage.