missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Sealing films for microplates and PCR plates
Premium sealing films for microplates, deepwell plates and PCR plates
Supplier: BRANDTECH™ BRAND™ 701392ES
Description
- Polyester film is optimized for ELISA applications, and is available plain, with colored strips for plate tracking, or in strips for easy well access
- Vinyl film that is piereceable/resealable for use in automation applications.
- Polyolefin film is highly transparent and suitable for real time qPCR and protein crystallization. It features a special non-tacky surface for easy placement and reduced chance of sample contaimination.
Chemical Identifiers
| Clear | |
| Not Autoclavable | |
| All well shapes |
| Non-sterile | |
| Pressure Sensitive |
Specifications
| Clear | |
| Polyester | |
| Not Autoclavable | |
| 5 Packs of 10 Sheets | |
| no | |
| Pressure Sensitive | |
| No | |
| All well shapes |
| DMSO Resistant | |
| Non-sterile | |
| 96/384 | |
| ANSI/SLAS | |
| Sealing Film | |
| Yes | |
| -40°C to +120°C | |
| Deep Well Plate |