Learn More
Mtc Bio Sealing film, Aluminum membrane, 10 bags of 10 membranes
Catalog No. NC1761395
Encompass
This item has a minimum qty of 3 packs per supplier requirements. PureAmp™ Sealing Membranes are compatible with all standard PCR plates. Four types are available for PCR and qPCR applications; pierceable aluminum, standard transparent, optically clear sticky adhesive, and optically clear pressure-sensitive adhesive, all of which are guaranteed to keep evaporation rates below 5%. RNase, DNase, DNA, and PCR inhibitor free Evaporation rates 5% Inner packs of 10 membranes
Catalog No. NC1761395 Supplier Mtc Bio Supplier No. P1001A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This item has a minimum qty of 3 packs per supplier requirements. PureAmp™ Sealing Membranes are compatible with all standard PCR plates. Four types are available for PCR and qPCR applications; pierceable aluminum, standard transparent, optically clear sticky adhesive, and optically clear pressure-sensitive adhesive, all of which are guaranteed to keep evaporation rates below 5%. RNase, DNase, DNA, and PCR inhibitor free Evaporation rates 5% Inner packs of 10 membranes