Learn More
Heidolph™ Seal (Gearhead Axis) for Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. 13900943
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator Drive Head
Catalog No. For Use With (Equipment)
13900943 Laborota 20, Hei-VAP Industrial Rotary Evaporator Drive Head
1 options
Catalog No. 13900943 Supplier Heidolph™ Supplier No. 036269400
Seal (gearhead axis) prevents the medium from reaching the ball bearings of the gearhead.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator
Specifications
Type Seal
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator Drive Head