Learn More
SeaKem™ LE Agarose, Lonza™ DFS Item
Ideal for routine analysis of nucleic acids by gel electrophoresis and blotting. SeaKem™ LE Agarose sharply resolves DNA and provides consistent resolution from lot-to-lot.
$2452.21 - $2452.21
Specifications
Chemical Name or Material SeaKem LE Agarose
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BMA50004 DFS Item
View Documents
Lonza Walkersville Inc
50004
500 g
Each for $2,452.21
Only null left
Description
Description
  • For DNA analysis/blotting
  • Molecular Biology Grade
  • EEO 0.09–0.13
  • Gelling temperature 36° ± 1.5°C
  • DNase/RNase-free
Specifications
Specifications
SeaKem LE Agarose