missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ Seahorse XF HS Mini Analyzer

Catalog No. S7852A
Change view
Click to view available options
Detection Method:
Fluorescence
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
S7852A Fluorescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7852A Supplier Agilent Technologies™ Supplier No. S7852A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seahorse XF HS Mini Analyzer provides real-time functional metabolic measurements in live cells, in an eight-well miniplate format. This entry-level analyzer delivers improved precision, Windows 10 compliance, and compatibility with Agilent™'s highest sensitivity miniplate.

  • Ideal for when biological material is limited
  • Multiple parameters reported from each assay well, including OCR, PER or ECAR, and ATP product rates
  • High OCR precision at low rates, providing confidence when working with quiescent cell types, such as naive T cells
  • High sensitivity that requires 3X fewer cells to achieve robust response
  • Most suited for studies with limited biological materials when working together with XF HS miniplates
  • Four-port injection system with automated mixing feature for assessing immediate cellular responses to substrates, inhibitors, and other compounds in real time
  • Wide operation environment temperature of 4°C to 30°C, allowing the analyzer to maintain internal assay temperature of 16°C to 42°C
  • Web-based analytics tool, Seahorse Analytics, for streamlined data analysis

Specifications

Type Cell Analyzer
Shaking No
For Use With (Application) Cell Metabolism and Function, Bioenergetics, Mitochondrial Health, Toxicity, Glycolysis, ATP Production Rate
Detection Method Fluorescence
Light Source LED
Voltage 100/240 V