missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Se-methylseleno-L-cysteine, 98%

CAS: 26046-90-2 | C4H9NO2Se | 182.092 g/mol

$169.82 - $427.23

Chemical Identifiers

CAS 26046-90-2
Molecular Formula C4H9NO2Se
Molecular Weight (g/mol) 182.092
MDL Number MFCD00800565
InChI Key XDSSPSLGNGIIHP-VKHMYHEASA-N
Synonym methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno
PubChem CID 147004
ChEBI CHEBI:27812
IUPAC Name (2R)-2-amino-3-methylselanylpropanoic acid
SMILES C[Se]CC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC300640500
SDS View Documents
Thermo Scientific Chemicals
300640500
50 mg Glass Bottle
Each for $169.82
Only null left
 
AC300642500
SDS View Documents
Thermo Scientific Chemicals
300642500
250 mg Glass Bottle
Each for $427.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

26046-90-2
182.092
XDSSPSLGNGIIHP-VKHMYHEASA-N
147004
(2R)-2-amino-3-methylselanylpropanoic acid
C4H9NO2Se
MFCD00800565
methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno
CHEBI:27812
C[Se]CC(C(=O)O)N

Specifications

26046-90-2
Authentic
C4H9NO2Se
MFCD00800565
XDSSPSLGNGIIHP-VKHMYHEASA-N
(2R)-2-amino-3-methylselanylpropanoic acid
182.092
CHEBI:27812
≥97.5%
Se-methylseleno-L-cysteine
White to Yellow
98%
Glass Bottle
methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno
C[Se]CC(C(=O)O)N
50 mg
147004
182.08
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only