missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Se-methylseleno-L-cysteine, 98%

Catalog No. AC300640500
Change view
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300640500 50 mg Glass Bottle
AC300642500 250 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC300640500 Supplier Thermo Scientific Chemicals Supplier No. 300640500
Only null left

CAS: 26046-90-2 | C4H9NO2Se | 182.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26046-90-2
Molecular Formula C4H9NO2Se
Molecular Weight (g/mol) 182.092
MDL Number MFCD00800565
InChI Key XDSSPSLGNGIIHP-VKHMYHEASA-N
Synonym methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno
PubChem CID 147004
ChEBI CHEBI:27812
IUPAC Name (2R)-2-amino-3-methylselanylpropanoic acid
SMILES C[Se]CC(C(=O)O)N

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 50 mg
Formula Weight 182.08
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Se-methylseleno-L-cysteine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only