Learn More
Fisherbrand™ Scullery Sink Faucet
Catalog No. 15050332
Change view
Click to view available options
Material:
Brass
Catalog No. Material
15050332 Brass
1 options
Catalog No. 15050332
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Scullery Sink Faucets have a splash mounted heavy duty construction and have lever style handles with a 12 in. (30.5 cm) spout.
  • Scullery Sink Faucets have a splash mounted heavy duty construction and have lever style handles with a 12 in. (30.5 cm) spout.
  • They are designed for a two hole, 8 in. (20.3 cm) on center mounting pattern and have 1/2 in. (1.3 cm) NPT female inlets.
Specifications
Material Brass
Length (English) Spout 12 in.
Length (Metric) Spout 30.5 cm