Learn More
ITL BioMedical SCU NARROW CAP SAMPLER
Catalog No. 22500126
Encompass
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
22500126 Sterile
1 options
Catalog No. 22500126 Supplier ITL BioMedical Supplier No. A100724
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The SCU Narrow Cap Sampler is a needle-free, single-use device for safe and efficient blood culture sampling. It supports drop and syringe methods, is compatible with narrow-neck and BD-style bottles, and ensures safety and flexibility in clinical lab settings.
  • Needle-free design eliminates sharps risk and reduces exposure
  • Dual-mode sampling: compatible with both drop and syringe methods
  • Narrow-cap design provides secure fit on most long-neck culture bottles
  • Protective wide cap minimizes contamination and ensures safer disposal
  • Broad compatibility with major blood culture bottle brands
  • Cost-effective alternative to exposed needle systems
Specifications
Material Plastic
Needle Gauge None
Latex Free Yes
Product Type Cap Sampler
Disposable Single-use
For Use With (Equipment) Narrow Neck Bottles, BD-Style Bottles
Connection Type Female Luer Lock
Graduated No
Sterility Sterile