Learn More
Brands SCRW CAP AND CLEAR VL KT 100
Catalog No. 50929261
Encompass
2mL Wide opening (9 mm) Screw Top Vials and Packs. The screw caps and precision fit septa give a secure seal with the microvolume inserts. Vials are 2mL, 12x32 mm, avaialbe in pre assembled vial packs and convenience packs for ease of use and ordering
Catalog No. 50929261 Supplier Brands Supplier No. 51820553
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
2mL Wide opening (9 mm) Screw Top Vials and Packs. The screw caps and precision fit septa give a secure seal with the microvolume inserts. Vials are 2mL, 12x32 mm, avaialbe in pre assembled vial packs and convenience packs for ease of use and ordering