missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ Scrubbing Towels

Catalog No. 19043742
Change view
Click to view available options
Quantity:
170 Wipes/Canister
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19043742 170 Wipes/Canister
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19043742 Supplier GOJO™ Supplier No. 6398 02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No water needed towels for use anywhere heavy duty cleaning is required. Dual textured wet towel for quick and convienient cleaning.

  • Dual textured with a scrubbing side for removing tough soils and a smooth side for quick dirt absorption
  • Extra-large towels for superior cleaning capacity
  • Contains skin conditioners to help prevent dryness and irritation
  • For use anywhere heavy duty cleaning is required
  • No water needed
  • Fresh citrus scent
  • Effective on oil, grease, paint, adhesives, tar, and asphalt

Specifications

Product Type Heavy Duty Hand Cleanser
Color Gray
Form Wipe
For Use With (Application) Effective on oil, grease, paint, adhesives, tar, and asphalt.
For Use With (Equipment) 6302-01
Packaging Bucket
Quantity 170 Wipes/Canister
Size 170 count