missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOJO™ Scrubbing Towels
Description
- Dual textured with a scrubbing side for removing tough soils and a smooth side for quick dirt absorption
- Extra-large towels for superior cleaning capacity
- Contains skin conditioners to help prevent dryness and irritation
- For use anywhere heavy duty cleaning is required
- No water needed
- Fresh citrus scent
- Effective on oil, grease, paint, adhesives, tar, and asphalt
- Packaged in a countertop display
Specifications
Specifications
| Product Type | Scrubbing Towel |
| Color | Gray |
| Description | Individually packaged |
| Packaging Quantity | 4/Cs. |
| Sterility | Non-sterile |