The CELLSCRIPT ScriptCap™ 2'-O-Methyltransferase Kit provides an enzymatic solution for converting Cap 0-RNA to Cap 1-RNA, enhancing both stability and translation efficiency. This kit achieves nearly 100% capping efficiency and simplifies the workflow by allowing direct input of capped RNA into further reactions without cleanup steps.
- Converts Cap 0-RNA to Cap 1-RNA
- ~100% capping efficiency
- Up to 50% increase in mRNA translation efficiency
- Reaction capacity of 60 g Cap 0-RNA per reaction
- 25 reactions per kit
- Storage at -20C
- Enzyme concentration: 100 U/l
- 30 min reaction incubation at 37C
- No cleanup required before poly(A) tailing
- For research use only