Learn More
Microsolv Technology Screwcap For 12X32 1000/Cs DFS Item
Supplier:  Microsolv Technology Corporation 9502S09PEK
Caps, Screw Top, Solid, Single injection use with a thinned penetration point. No Septa required. LCMS compatible, knurled polyethylene, Natural color Caps. For use with with 9-425mm thread, 12x32mm autosampler vials. AQ Brand. 1000/CS.
Catalog No. NC1309328
Only null left
Explore available promotions