missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Screw Cap Septum Vials

For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials.

Supplier:  Thermo Scientific™ TS13028

 View more versions of this product

Catalog No. PI13028


Only null left

Description

Description

For a convenient, versatile, and cost-effective vial and closure system, choose Thermo Scientific™ Screw Cap Septum Vials. Made of autoclavable borosilicate glass and available with a wide assortment of special closures and accessories, these vials are ideal for sample collection and small derivatization reactions for use with automated gas (GC) and liquid chromatography (LC), and for storage of reagents and standards under complete seal with instant syringe access. The heavy-duty, flip-top divider box provides easy access to vials, caps, and septa and offers a convenient sample storage center. Clear and amber vials are available.

Applications

The 40mL clear or amber vial with a Thermo Scientific Disc is suitable for discrete water sampling under U.S. EPA 40 CFR Parts 136 and 141.

The 1.5mL clear and amber vials fit GC and LC autosamplers that use standard 12 ́ 32 mm vials.

Specifications

Chemical Identifiers

Borosilicate Glass
Screw Cap
Autoclavable
Septum Vial

Specifications

7mL; Clear
17 mm
7 mL
Screw Cap
Autoclavable
60 mm
Clear
Borosilicate Glass
7 mL
15-425
Certificate of Analysis (COA), Certificate of Conformance (COC)
Septum Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.