missing translation for 'onlineSavingsMsg'
Learn More

Scott's Bluing Reagent (Scott's Tapwater Substitute), Bluing Agent for Nuclear Hematoxylin Stain, Ricca Chemical

Catalog No. 669732
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
669732 1 L Natural Poly Bottle
66971 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 669732 Supplier Ricca Chemical Company Supplier No. 669732
Only null left

Bluing Agent for Nuclear Hematoxylin Stain

Specifications

Physical Form Liquid
Chemical Name or Material Scott's Bluing Reagent
Name Note Scott's Tapwater Substitute
CAS 7732-18-5, 10034-99-8, 144-55-8, 50-00-0, 67-56-1
CAS Min % 95.0, 1.91, 0.51, 0.19
CAS Max % 99.0, 1.99, 0.53, 0.2
Packaging Natural Poly Bottle
Color Colorless
Quantity 1 L
Grade Laboratory