missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scott's Bluing Reagent (Scott's Tapwater Substitute), Bluing Agent for Nuclear Hematoxylin Stain, Ricca Chemical
Specifications
Specifications
| Physical Form | Solution |
| CAS | 7732-18-5, 10034-99-8, 144-55-8, 50-00-0, 67-56-1 |
| CAS Min % | 95.0, 1.91, 0.51, 0.19 |
| CAS Max % | 99.0, 1.99, 0.53, 0.2 |
| Packaging | Natural Poly Bottle |
| Color | Blue |
| Quantity | 4 L |
| Grade | Laboratory |