Learn More
3M™ Scotch™ Paint Masking Tape, 231/231A
High-performance crepe paper masking tape designed to perform well in most industrial painting situations, including the use of bake cycles.
Supplier:  3M™ 7000088514
Catalog No. 296360300
Only null left
Description
Description
  • Rubber adhesive tape with instant adhesion that sticks easily and allows fast application
  • It features good holding power that resists lifting or curling to help reduce rework and superior adhesive transfer resistance with clean removal
  • Typical physical and performance characteristics include: thickness: 7.6 mil, adhesion to steel: 38 oz./in. width, tensile strength: 28 lbs/in width, elongation at break: 10%, temperature use range: up to 300°F (148°C)
  • The smooth mini-crepe backing results in excellent surface contact with very secure holding
  • It provides excellent conformability and easy handling that helps reduce time involved in application
  • The backing is solvent and moisture-resistant to help reduce bleed-through and has good dead stretch for minimal recovery
  • It is ideal for easy-to-tear backing aids in handling, yet strong enough to resist slivering, giving one-piece removal
  • It is paint flake resistant to help reduce rework due to contamination
  • It can be certified for ASTM-D6123/D6123M97 and it replaces obsolete PPP-T-42C, Type 1, (CID-A-A-883A).
Specifications
Specifications
Rubber
36
60.14 yd.
24 mm
Tan
24 mm x 55 m
0.94 in.
Product Suggestions
Product Suggestions