Learn More
3M™ Scotch™ Paint Masking Tape, 231
Catalog No. 296360301
Change view
Click to view available options
Color:
Tan
Catalog No. Color
296360301 Tan
1 options
Catalog No. 296360301 Supplier 3M™ Supplier No. 7000088515
High-temperature resistant tape for use in masking and holding applications
  • Highly cured rubber adhesive for excellent high-temperature holding and adhesive transfer resistance to keep edge lifting minimized to help reduce rework and labor
  • It features specially treated crepe paper backing that is sliver resistant, conformable, easy to tear, and solvent and moisture resistant
  • It is designed for indoor use only as it should not be subjected to outdoor exposure
  • It is ideal for use on aluminum anodized surfaces
Specifications
Adhesive Type Rubber
Color Tan
Quantity 24/Cs.
Dimensions (L x W) 36 mm x 55 m
Length (English) 60.14 yd.
Width (English) 1.41 in.
Width (Metric) 36 mm