missing translation for 'onlineSavingsMsg'
Learn More

3M™ Scotch-Brite™ Power Sponge 3000

Catalog No. 17623927
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17623927 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17623927 Supplier 3M™ Supplier No. 7100064740

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Scotch-Brite™ Power Sponge 3000 - Bow tie shaped dual action scouring sponge.

  • Scotch-Brite™ Power Pad combined with a sponge.
  • Antimicrobial protection is locked-in to resist odors in the sponge.
  • Up to 8 times less abrasive than other medium duty commercial scouring products

Specifications

Product Type Scrub Sponge
Color Blue
For Use With (Application) Commercial Kitchen Cleaning
Length (English) 4.5 in.
Length (Metric) 114.3 mm
Quantity 20/Cs.
Width (English) 2.8 in.
Width (Metric) 71.12 mm