missing translation for 'onlineSavingsMsg'
Learn More

Scopolamine Hydrobromide Trihydrate 98.0+%, TCI America™

Catalog No. S00211G
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00211G 1 g
S002110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S00211G Supplier TCI America Supplier No. S00211G
Only null left

Chemical Identifiers

CAS 6533-68-2
Molecular Formula C17H28BrNO7
Molecular Weight (g/mol) 438.32
MDL Number MFCD00150066
InChI Key LACQPOBCQQPVIT-FCFGRTKISA-N
Synonym hyoscine hydrobromide trihydrate
PubChem CID 131632304
IUPAC Name hydrogen (1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate trihydrate bromide
SMILES [H+].O.O.O.[Br-].CN1[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 1 g
UN Number 1544
Formula Weight 384.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Scopolamine Hydrobromide Trihydrate
RTECSNumber YM4550000
TSCA Yes